Thermo Scientific Chemicals

Isovaleric acid, 98%, Thermo Scientific Chemicals

Catalog number: A18642.AK
250 mL, Each
Thermo Scientific Chemicals

Isovaleric acid, 98%, Thermo Scientific Chemicals

Catalog number: A18642.AK
250 mL, Each
Quantity
Catalog number: A18642.AK
also known as A18642-AK
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
503-74-2
IUPAC Name
3-methylbutanoic acid
Molecular Formula
C5H10O2
InChI Key
GWYFCOCPABKNJV-UHFFFAOYSA-N
SMILES
CC(C)CC(O)=O
Appearance (Color)
Clear colorless
Assay (Aqueous acid-base Titration)
≥97.5 to ≤102.5%
Assay (GC)
≥97.5%
Identification (FTIR)
Conforms
Refractive Index
1.4010-1.4050 @ 20?C

Description

A molecular entity capable of donating a hydron to an acceptor (Bronsted base). It is a favorable carbon source for cell growth. Moreover, it is a promising odor indicator.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
A molecular entity capable of donating a hydron to an acceptor (Bronsted base). It is a favorable carbon source for cell growth. Moreover, it is a promising odor indicator.

Solubility
Soluble in water 20°C 25 g/L. Soluble in ethanol, chloroform, ether.

Notes
Store away from oxidizing agents and bases. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text