Thermo Scientific Chemicals

4'-Bromo-2'-chloroacetanilide, 98+%, Thermo Scientific Chemicals

Catalog number: A18652.22
100 g, Each
Thermo Scientific Chemicals

4'-Bromo-2'-chloroacetanilide, 98+%, Thermo Scientific Chemicals

Catalog number: A18652.22
100 g, Each
Quantity
Catalog number: A18652.22
also known as A18652-22
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
3460-23-9
IUPAC Name
N-(4-bromo-2-chlorophenyl)acetamide
Molecular Formula
C8H7BrClNO
InChI Key
MITWNEIUIPGZKR-UHFFFAOYSA-N
SMILES
CC(=O)NC1=CC=C(Br)C=C1Cl
Appearance (Color)
White to cream
Form
Crystals or powder or crystalline powder
Assay (GC)
≥98.0%
Melting Point (clear melt)
150.0-157.0°C

Description

It is employed as a intermediate for pharmaceutical.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is employed as a intermediate for pharmaceutical.

Solubility
Soluble in hot methanol (very faint turbidity).

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text