Sodium cyclamate, 98%, Thermo Scientific Chemicals
Sodium cyclamate, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Sodium cyclamate, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5000 g
250 g
Catalog number A18666.0I
also known as A18666-0I
Price (USD)/ Each
520.65
Online exclusive
578.00 
Save 57.35 (10%)
-
Add to cart
Quantity:
5000 g
Request bulk or custom format
Price (USD)/ Each
520.65
Online exclusive
578.00 
Save 57.35 (10%)
Add to cart
Sodium cyclamate, 98%, Thermo Scientific Chemicals
Catalog numberA18666.0I
Price (USD)/ Each
520.65
Online exclusive
578.00 
Save 57.35 (10%)
-
Add to cart
Chemical Identifiers
CAS139-05-9
IUPAC Namesodium N-cyclohexylsulfamate
Molecular FormulaC6H12NNaO3S
InChI KeyUDIPTWFVPPPURJ-UHFFFAOYSA-M
SMILES[Na+].[O-]S(=O)(=O)NC1CCCCC1
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (Non-aqueous acid-base Titration)≥97.5 to ≤102.5%
Appearance (Color)White
FormCrystals or powder or crystalline powder or fused solid
Sodium Cyclamate is an artificial sweetener and is 30 times as saccharose's. It is widely used in pickles, seasoning sauce, cakes, biscuits, bread, ice cream, frozen sucker, popsicles, drinks and so on, with a maximum amount of 0.65g/kg. Secondly, it is used in confect, with a maximum amount of 1.0g/kg. Thirdly, it is used in orange peel, preserved plum, dried arbutus and so on, with the largest amount of 8.0g/kg. It is also used in cosmetics and pharmaceutical industry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Sodium Cyclamate is an artificial sweetener and is 30 times as saccharose′s. It is widely used in pickles, seasoning sauce, cakes, biscuits, bread, ice cream, frozen sucker, popsicles, drinks and so on, with a maximum amount of 0.65g/kg. Secondly, it is used in confect, with a maximum amount of 1.0g/kg. Thirdly, it is used in orange peel, preserved plum, dried arbutus and so on, with the largest amount of 8.0g/kg. It is also used in cosmetics and pharmaceutical industry.

Solubility
Soluble in water (>=10 g/100 mL at 20°C).

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
RUO – Research Use Only

General References:

  1. F.J.C. Roe.; L.S. Levy.; R.L. Carter. Feeding studies on sodium cyclamate, saccharin and sucrose for carcinogenic and tumour-promoting activity.Food and Cosmetics Toxicology,1970,8(2), 135-145.
  2. Thomas A. Biemer. Analysis of saccharin, acesulfame-K and sodium cyclamate by high-performance ion chromatography.J. Chromatogr. A,1989,463463-468.