Sulfamide, 99%, Thermo Scientific Chemicals
Sulfamide, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Sulfamide, 99%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
2.5 g
50 g
Catalog number A18680.09
also known as A18680-09
Price (USD)/ Each
105.00
-
Add to cart
Quantity:
10 g
Request bulk or custom format
Price (USD)/ Each
105.00
Add to cart
Sulfamide, 99%, Thermo Scientific Chemicals
Catalog numberA18680.09
Price (USD)/ Each
105.00
-
Add to cart
Chemical Identifiers
CAS7803-58-9
IUPAC Namesulfamoylamine
Molecular FormulaH4N2O2S
InChI KeyNVBFHJWHLNUMCV-UHFFFAOYSA-N
SMILESNS(N)(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
FormCrystalline powder
Melting Point89-93?C
Assay from Supplier's CofA≥98.5%
Appearance (Color)White
It is employed as a carbonic anhydrase inhibitor. It is widely used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is employed as a carbonic anhydrase inhibitor. It is widely used as a pharmaceutical intermediate.

Solubility
Soluble in water, hot alcohol, acetone. Sparingly soluble in cold alcohol

Notes
Moisture Sensitive. Store away from oxidizing agents, water/humidity. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
RUO – Research Use Only

General References:

  1. Deborah Sterling.; Reinhart A. F. Reithmeier.; and Joseph R. Casey. A Transport Metabolon. Functional Interaction Of Carbonic Anhydrase Ii And Chloride/Bicarbonate Exchangers. J. Bio. Chem.,2001,27647886-47894.
  2. Maryanoff BE.; McComsey DF.; Costanzo MJ.; Hochman C.; Smith-Swintosky V.; Shank RP. Comparison of sulfamate and sulfamide groups for the inhibition of carbonic anhydrase-II by using topiramate as a structural platform.J Med Chem.2005,48(6), 1941-1947.