Thermo Scientific Chemicals

Sulfamide, 99%, Thermo Scientific Chemicals

Catalog number: A18680.18
50 g, Each
Thermo Scientific Chemicals

Sulfamide, 99%, Thermo Scientific Chemicals

Catalog number: A18680.18
50 g, Each
Quantity
Catalog number: A18680.18
also known as A18680-18
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
7803-58-9
IUPAC Name
sulfamoylamine
Molecular Formula
H4N2O2S
InChI Key
NVBFHJWHLNUMCV-UHFFFAOYSA-N
SMILES
NS(N)(=O)=O
Form
Crystalline powder
Melting Point
89-93?C
Assay from Supplier's CofA
≥98.5%
Appearance (Color)
White

Description

It is employed as a carbonic anhydrase inhibitor. It is widely used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is employed as a carbonic anhydrase inhibitor. It is widely used as a pharmaceutical intermediate.

Solubility
Soluble in water, hot alcohol, acetone. Sparingly soluble in cold alcohol

Notes
Moisture Sensitive. Store away from oxidizing agents, water/humidity. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text