1-Bromo-2-naphthol, 97%, Thermo Scientific Chemicals
1-Bromo-2-naphthol, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

1-Bromo-2-naphthol, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Catalog number A18692.14
also known as A18692-14
Price (USD)/ Each
57.50
-
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (USD)/ Each
57.50
Add to cart
1-Bromo-2-naphthol, 97%, Thermo Scientific Chemicals
Catalog numberA18692.14
Price (USD)/ Each
57.50
-
Add to cart
Chemical Identifiers
CAS573-97-7
IUPAC Name1-bromonaphthalen-2-ol
Molecular FormulaC10H7BrO
InChI KeyFQJZPYXGPYJJIH-UHFFFAOYSA-N
SMILESOC1=CC=C2C=CC=CC2=C1Br
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Cream to pale brown
Assay (GC)≥96.0%
FormPowder
Melting Point (clear melt)73.0-83.0?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only