Thermo Scientific Chemicals

1,4-Benzodioxane-6-carboxaldehyde, 99%, Thermo Scientific Chemicals

Catalog number: A18696.06
5 g, Each
Thermo Scientific Chemicals

1,4-Benzodioxane-6-carboxaldehyde, 99%, Thermo Scientific Chemicals

Catalog number: A18696.06
5 g, Each
Quantity
Catalog number: A18696.06
also known as A18696-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
29668-44-8
IUPAC Name
2,3-dihydro-1,4-benzodioxine-6-carbaldehyde
Molecular Formula
C9H8O3
InChI Key
CWKXDPPQCVWXAG-UHFFFAOYSA-N
SMILES
O=CC1=CC=C2OCCOC2=C1
Appearance (Color)
Pale cream to pale brown
Form
Crystals or powder or crystalline powder
Assay (GC)
≥98.5%
Melting Point (clear melt)
47.0-54.0?C

Description

1,4-Benzodioxan-6-carboxaldehyde finds its uses as building block in the synthesis of tetrahydroisoquinolinones, in the preparation of benzofuran analog, a potential inhibitor of CAMP-specific phosphodiesterase type IV, in the synthesis of (5Z)-5-(2,3-dihydro-1,4-benzodioxan-6-ylmethylene)-1-methyl-2-thioxoimidazolidin-4-one. It was screened as a fragment for inhibitors of PDEA using enthalpy arrays and X-ray crystallography. 1,4-Benzodioxane-6-carboxaldehyde have also been used as an intermediate in the synthesis of a new class of Dopamine D3 and D4 receptor antagonists.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,4-Benzodioxan-6-carboxaldehyde finds its uses as building block in the synthesis of tetrahydroisoquinolinones, in the preparation of benzofuran analog, a potential inhibitor of CAMP-specific phosphodiesterase type IV, in the synthesis of (5Z)-5-(2,3-dihydro-1,4-benzodioxan-6-ylmethylene)-1-methyl-2-thioxoimidazolidin-4-one. It was screened as a fragment for inhibitors of PDEA using enthalpy arrays and X-ray crystallography. 1,4-Benzodioxane-6-carboxaldehyde have also been used as an intermediate in the synthesis of a new class of Dopamine D3 and D4 receptor antagonists.

Solubility
Solubility in methanol is almost transparent.

Notes
Store away from air, oxidizing agents and bases. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text