Thermo Scientific Chemicals

2,2,2-Tribromoethanol, 99%, Thermo Scientific Chemicals

Catalog number: A18706.14
25 g, Each
Thermo Scientific Chemicals

2,2,2-Tribromoethanol, 99%, Thermo Scientific Chemicals

Catalog number: A18706.14
25 g, Each
Quantity
Catalog number: A18706.14
also known as A18706-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
75-80-9
IUPAC Name
2,2,2-tribromoethan-1-ol
Molecular Formula
C2H3Br3O
InChI Key
YFDSDPIBEUFTMI-UHFFFAOYSA-N
SMILES
OCC(Br)(Br)Br
Form
Crystalline powder or crystals or powder and/or lumps
Water Content (Karl Fischer Titration)
<1.0%
Appearance (Color)
White to cream to pale brown
Assay (GC)
≥98.5%

Description

2,2,2-Tribromoethanol is utilized in organic synthesis of beta-amino alcohols. The pharmaceutical preparation of tribromoethanol serves as an anesthetic in medicine.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,2,2-Tribromoethanol is utilized in organic synthesis of beta-amino alcohols. The pharmaceutical preparation of tribromoethanol serves as an anesthetic in medicine.

Solubility
Soluble in alcohol, ether, benzene and warmpetroleum ether. Partially soluble in water.

Notes
Light Sensitive. Store in cool and well ventilated area. Keep container tightly closed.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text