N,N'-Diphenylurea, 98%, Thermo Scientific Chemicals
N,N'-Diphenylurea, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

N,N'-Diphenylurea, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
25 g
Catalog number A18720.22
also known as A18720-22
Price (USD)/ Each
125.00
-
Add to cart
Quantity:
100 g
Request bulk or custom format
Price (USD)/ Each
125.00
Add to cart
N,N'-Diphenylurea, 98%, Thermo Scientific Chemicals
Catalog numberA18720.22
Price (USD)/ Each
125.00
-
Add to cart
Chemical Identifiers
CAS102-07-8
SpecificationsSpecification SheetSpecification Sheet
Melting Point (clear melt)237.0-243.0?C
Identification (FTIR)Conforms
Appearance (Color)White to pale brown
FormCrystals or powder or crystalline powder
Assay (HPLC)≥97.5%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only