It finds use in artificial oil of bergamot, myrrh and lemon, and is an excellent fixative for neroli bases.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
It finds use in artificial oil of bergamot, myrrh and lemon, and is an excellent fixative for neroli bases.
Solubility
Soluble in alcohol, water, (0.008994 mg/L @ 25°C (est)).
Notes
Air Sensitive. Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
RUO – Research Use Only
Nanda K. Gulavita, et al. Nitrogenous bisabolene sesquiterpenes from marine invertebrates.J. Org. Chem.,1986,51(26), 5136-5139.
Jae-Ha Ryu.; Yeon Su Jeong.; and Dong Hwan Sohn. A New Bisabolene Epoxide from Tussilago farfara, and Inhibition of Nitric Oxide Synthesis in LPS-Activated Macrophages.J. Nat. Prod.,1999,62(10), 1437-1438.