Thermo Scientific Chemicals

Bisabolene, mixture of isomers, Thermo Scientific Chemicals

Catalog number: A18724.18
50 g, Each
Thermo Scientific Chemicals

Bisabolene, mixture of isomers, Thermo Scientific Chemicals

Catalog number: A18724.18
50 g, Each
Quantity
Catalog number: A18724.18
also known as A18724-18
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
495-62-5
IUPAC Name
(4Z)-1-methyl-4-(6-methylhept-5-en-2-ylidene)cyclohex-1-ene
Molecular Formula
C15H24
InChI Key
XBGUIVFBMBVUEG-CCEZHUSRSA-N
SMILES
CC(C)=CCCC(C)=C1CCC(C)=CC1
Appearance (Color)
Clear colorless to pale yellow
Refractive Index
1.4850-1.5050 @ 20?C
Specific Gravity
0.845-0.865
Form
Liquid

Description

It finds use in artificial oil of bergamot, myrrh and lemon, and is an excellent fixative for neroli bases.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It finds use in artificial oil of bergamot, myrrh and lemon, and is an excellent fixative for neroli bases.

Solubility
Soluble in alcohol, water, (0.008994 mg/L @ 25°C (est)).

Notes
Air Sensitive. Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text