3',4'-Dichloropropiophenone, 98%, Thermo Scientific Chemicals
3',4'-Dichloropropiophenone, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

3',4'-Dichloropropiophenone, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
Catalog number A18740.14
also known as A18740-14
Price (USD)/ Each
199.00
-
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (USD)/ Each
199.00
Add to cart
3',4'-Dichloropropiophenone, 98%, Thermo Scientific Chemicals
Catalog numberA18740.14
Price (USD)/ Each
199.00
-
Add to cart
Chemical Identifiers
CAS6582-42-9
IUPAC Name1-(3,4-dichlorophenyl)propan-2-one
Molecular FormulaC9H8Cl2O
InChI KeyAJUAASWQUWIMHM-UHFFFAOYSA-N
SMILESCC(=O)CC1=CC=C(Cl)C(Cl)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to pale cream
FormCrystals or crystalline powder or fused solid
Assay (GC)≥97.5%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only