2-Amino-4-ethylpyridine is used as a precursor to prepare 1-ethyl-3-[5-(7-ethyl-imidazo[1,2-a]pyridin-2-yl)-2-hydroxy-phenyl]-urea. It is also used as an intermediate in organic synthesis. It is considered as an analogue of pirlimycin, which acts as an antibiotic.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
2-Amino-4-ethylpyridine is used as a precursor to prepare 1-ethyl-3-[5-(7-ethyl-imidazo[1,2-a]pyridin-2-yl)-2-hydroxy-phenyl]-urea. It is also used as an intermediate in organic synthesis. It is considered as an analogue of pirlimycin, which acts as an antibiotic.
Solubility
Soluble in water, hexane, ethyl acetate and methyl tert-butyl ether.
Notes
Incompatible with strong oxidizing agents, strong acids and bases. Store in a cool place.
RUO – Research Use Only
Qi, Z. C.; Han, S. Y.; Wu, Z. Y.; Chen, F. Y.; Cao, X. W.; Lian, H. Z.; Mao, L. Retention Prediction and Hydrophobicity Measurement of Weakly Basic Compounds in Reversed-phase Liquid Chromatography Using Ammonia and Triethylamine as Ion-suppressors. Curr. Anal. Chem. 2014, 10 (1), 172-181.
Ferrera-Lorenzo, N.; Fuente, E.; Bermúdez, J. M.; Suárez-Ruiz, I.; Ruiz, B. Conventional and microwave pyrolysis of a macroalgae waste from the Agar-Agar industry. Prospects for bio-fuel production. Bioresour. Technol. 2014, 151, 199-206.