Thermo Scientific Chemicals

2-Amino-4-ethylpyridine, 97%, Thermo Scientific Chemicals

Catalog number: A18754.88
2.5 g, Each
Thermo Scientific Chemicals

2-Amino-4-ethylpyridine, 97%, Thermo Scientific Chemicals

Catalog number: A18754.88
2.5 g, Each
Quantity
Catalog number: A18754.88
also known as A18754-88
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
33252-32-3
Appearance (Color)
White to pale cream
Form
Crystals or powder or crystalline powder
Melting Point (clear melt)
63.5-69.5?C
Assay (GC)
≥96.0%

Description

2-Amino-4-ethylpyridine is used as a precursor to prepare 1-ethyl-3-[5-(7-ethyl-imidazo[1,2-a]pyridin-2-yl)-2-hydroxy-phenyl]-urea. It is also used as an intermediate in organic synthesis. It is considered as an analogue of pirlimycin, which acts as an antibiotic.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Amino-4-ethylpyridine is used as a precursor to prepare 1-ethyl-3-[5-(7-ethyl-imidazo[1,2-a]pyridin-2-yl)-2-hydroxy-phenyl]-urea. It is also used as an intermediate in organic synthesis. It is considered as an analogue of pirlimycin, which acts as an antibiotic.

Solubility
Soluble in water, hexane, ethyl acetate and methyl tert-butyl ether.

Notes
Incompatible with strong oxidizing agents, strong acids and bases. Store in a cool place.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text