Thermo Scientific Chemicals

Manganese(II) 2,4-pentanedionate, Thermo Scientific Chemicals

Catalog number: A18762.14
25 g, Each
Thermo Scientific Chemicals

Manganese(II) 2,4-pentanedionate, Thermo Scientific Chemicals

Catalog number: A18762.14
25 g, Each
Quantity
Catalog number: A18762.14
also known as A18762-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
14024-58-9
IUPAC Name
manganese(2+) bis((2Z)-4-oxopent-2-en-2-olate)
Molecular Formula
C10H14MnO4
InChI Key
ZQZQURFYFJBOCE-FDGPNNRMSA-L
SMILES
[Mn++].C\C([O-])=C\C(C)=O.C\C([O-])=C\C(C)=O
Elemental Analysis
Manganese Mn: 20.40 - 23.00% (Theory 21.70%)
Appearance (Color)
Pale brown to brown
Assay (unspecified)
≥94.0 to ≤106.0% by Mn EA
Form
Powder

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text