Oxindole is considered as indole analogue, which shows pharmacological activity.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Oxindole is considered as indole analogue, which shows pharmacological activity.
Solubility
Soluble in methanol, ethanol, dimethyl sulfoxide and ether. Insoluble in water.
Notes
Incompatible with strong oxidizing agents.
RUO – Research Use Only
General References:
- Yang, H. B.; Zhao, Y. Z.; Sang, R.; Shi, M. (DHQ)2AQN-Catalyzed Asymmetric Substitution of Isatin-Derived Hydrazones with O-Boc-Protected Morita-Baylis-Hillman Adducts: A Strategy for Synthesizing Enantioenriched Azo Compounds Incorporating an Oxindole Scaffold. J. Org. Chem. 2014, 79 (8), 3519-3528.
- Hinze, M. E.; Daughtry, J. L.; Lewis, C. A. Access to the Surugatoxin Alkaloids: Chemo-, Regio-, and Stereoselective Oxindole Annulation. J. Org. Chem. 2015, 80 (22), 11258-11265.