Thermo Scientific Chemicals

Oxindole, 97+%, Thermo Scientific Chemicals

Catalog number: A18764.14
25 g, Each
Thermo Scientific Chemicals

Oxindole, 97+%, Thermo Scientific Chemicals

Catalog number: A18764.14
25 g, Each
Quantity
Catalog number: A18764.14
also known as A18764-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
59-48-3
IUPAC Name
1H-indol-2-ol
Molecular Formula
C8H7NO
InChI Key
JHFAEUICJHBVHB-UHFFFAOYSA-N
SMILES
OC1=CC2=CC=CC=C2N1
Appearance (Color)
White to dark cream to cream to brown
Identification (FTIR)
Conforms
Form
Powder
Melting Point (clear melt)
120.0-130.0?C
Assay (GC)
≥97.0%

Description

Oxindole is considered as indole analogue, which shows pharmacological activity.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Oxindole is considered as indole analogue, which shows pharmacological activity.

Solubility
Soluble in methanol, ethanol, dimethyl sulfoxide and ether. Insoluble in water.

Notes
Incompatible with strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text