Thermo Scientific Chemicals

N,N-Dimethylbenzamide, 98+%, Thermo Scientific Chemicals

Catalog number: A18766.22
100 g, Each
Thermo Scientific Chemicals

N,N-Dimethylbenzamide, 98+%, Thermo Scientific Chemicals

Catalog number: A18766.22
100 g, Each
Quantity
Catalog number: A18766.22
also known as A18766-22
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
611-74-5
IUPAC Name
N,N-dimethylbenzamide
Molecular Formula
C9H11NO
InChI Key
IMNDHOCGZLYMRO-UHFFFAOYSA-N
SMILES
CN(C)C(=O)C1=CC=CC=C1
Assay (GC)
≥98.0%
Melting Point (clear melt)
38.0-46.0°C
Appearance (Color)
White to cream or pale yellow
Form
Crystals or powder or crystalline powder or lumps or chunks or fused solid
Identification (FTIR)
Conforms

Description

Reagent for deoxygenation of secondary alcohols.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Reagent for deoxygenation of secondary alcohols.

Solubility
Soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text