N,N-Dimethylbenzamide, 98+%, Thermo Scientific Chemicals
N,N-Dimethylbenzamide, 98+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

N,N-Dimethylbenzamide, 98+%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
25 g
100 g
Catalog number A18766.36
also known as A18766-36
Price (USD)/ Each
516.00
-
Add to cart
Quantity:
500 g
Request bulk or custom format
Price (USD)/ Each
516.00
Add to cart
N,N-Dimethylbenzamide, 98+%, Thermo Scientific Chemicals
Catalog numberA18766.36
Price (USD)/ Each
516.00
-
Add to cart
Chemical Identifiers
CAS611-74-5
IUPAC NameN,N-dimethylbenzamide
Molecular FormulaC9H11NO
InChI KeyIMNDHOCGZLYMRO-UHFFFAOYSA-N
SMILESCN(C)C(=O)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (GC)≥98.0%
Melting Point (clear melt)38.0-46.0°C
Appearance (Color)White to cream or pale yellow
FormCrystals or powder or crystalline powder or lumps or chunks or fused solid
Identification (FTIR)Conforms
Reagent for deoxygenation of secondary alcohols.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Reagent for deoxygenation of secondary alcohols.

Solubility
Soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
RUO – Research Use Only

General References:

  1. Ji Young Kim et. al. Hydrotropic polymer micelles as versatile vehicles for delivery of poorly water-soluble drugs. Journal of Controlled Release, 2011, 152 (1), 13-20.
  2. Yokoo K, et al. Reactions of ethyl-and phenyllanthanide s complexes with n, n-dimethylbenzamide and benzaldehyde. Polyhedron , 1983, 2(10), 1101-02.
  3. Has been used as an acylating agent for alkynyl boranes: Chem. Lett., 35 (1983).Similarly, reacts with Grignard reagents in ether, to give phenyl ketones in good yields: Synthesis, 228 (1984). For o-lithiation of N,N-dialkylbenzamides, see N,N-Diethyl benzamide, L08427 and N,N-Diisopropyl benzamide, B22918.