It was used in preparation of pyrokinin class of insect neuropeptides. It was also used in preparation of N-(9-fluoreneacetyl)-L-leucine.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
It was used in preparation of pyrokinin class of insect neuropeptides. It was also used in preparation of N-(9-fluoreneacetyl)-L-leucine.
Solubility
Soluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
RUO – Research Use Only
P E Teal; and R J Nachman. Prolonged pheromonotropic activity of pseudopeptide mimics of insect pyrokinin neuropeptides after topical application or injection into a moth. Regulatory Peptides, 1997, 72 (2-3), 161-167.
Isaac O Donkor; Haregewein Assefa; Jiuyu Liu. Structural basis for the potent calpain inhibitory activity of peptidyl alpha-ketoacids. Journal of medicinal and pharmaceutical chemistry, 2008, 51(14), 4346-4350.