Fluorene-9-acetic acid, 99.5+%, Thermo Scientific Chemicals
Fluorene-9-acetic acid, 99.5+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Fluorene-9-acetic acid, 99.5+%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
Catalog number A18772.06
also known as A18772-06
Price (USD)/ Each
250.00
-
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)/ Each
250.00
Add to cart
Fluorene-9-acetic acid, 99.5+%, Thermo Scientific Chemicals
Catalog numberA18772.06
Price (USD)/ Each
250.00
-
Add to cart
Chemical Identifiers
CAS6284-80-6
IUPAC Name2-(9H-fluoren-9-yl)acetic acid
Molecular FormulaC15H12O2
InChI KeyWFSMJMTYIMFHPV-UHFFFAOYSA-N
SMILESOC(=O)CC1C2=CC=CC=C2C2=CC=CC=C12
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (Aqueous acid-base Titration)≥99.0 to ≤101.0%
Melting Point (clear melt)134.5-140.5?C
FormCrystals or powder or crystalline powder
Assay (HPLC)≥99.0%
Appearance (Color)White to pale cream
It was used in preparation of pyrokinin class of insect neuropeptides. It was also used in preparation of N-(9-fluoreneacetyl)-L-leucine.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It was used in preparation of pyrokinin class of insect neuropeptides. It was also used in preparation of N-(9-fluoreneacetyl)-L-leucine.

Solubility
Soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
RUO – Research Use Only

General References:

  1. P E Teal; and R J Nachman. Prolonged pheromonotropic activity of pseudopeptide mimics of insect pyrokinin neuropeptides after topical application or injection into a moth. Regulatory Peptides, 1997, 72 (2-3), 161-167.
  2. Isaac O Donkor; Haregewein Assefa; Jiuyu Liu. Structural basis for the potent calpain inhibitory activity of peptidyl alpha-ketoacids. Journal of medicinal and pharmaceutical chemistry, 2008, 51(14), 4346-4350.