5-Fluoro-2-nitrobenzotrifluoride, 98+%, Thermo Scientific Chemicals
5-Fluoro-2-nitrobenzotrifluoride, 98+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

5-Fluoro-2-nitrobenzotrifluoride, 98+%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number A18780.06
also known as A18780-06
Price (USD)/ Each
53.60
-
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)/ Each
53.60
Add to cart
5-Fluoro-2-nitrobenzotrifluoride, 98+%, Thermo Scientific Chemicals
Catalog numberA18780.06
Price (USD)/ Each
53.60
-
Add to cart
Chemical Identifiers
CAS393-09-9
IUPAC Name4-fluoro-1-nitro-2-(trifluoromethyl)benzene
Molecular FormulaC7H3F4NO2
InChI KeyWMQOSURXFLBTPC-UHFFFAOYSA-N
SMILES[O-][N+](=O)C1=CC=C(F)C=C1C(F)(F)F
View more
SpecificationsSpecification SheetSpecification Sheet
FormLiquid
Refractive Index1.4580-1.4620 @ 20?C
Appearance (Color)Clear yellow
Assay (GC)≥98.0%
5-Fluoro-2-nitrobenzotrifluoride may be used as monomer in the synthesis of hyperbranched poly(aryl ether). Also used in the preparation of androgen receptor modulators and other pharmaceutical compounds.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
5-Fluoro-2-nitrobenzotrifluoride may be used as monomer in the synthesis of hyperbranched poly(aryl ether). Also used in the preparation of androgen receptor modulators and other pharmaceutical compounds.

Solubility
Soluble in chloroform and methanol.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
RUO – Research Use Only

General References:

  1. Dhara MG; and Banerjee S. Fluorinated high-performance polymers: Poly (arylene ether) s and aromatic polyimides containing trifluoromethyl groups. Prog. Polym. Sci., 2010, 35 (8), 1022-27.
  2. Trump, R.P. et al. Design and Synthesis of an Array of Selective Androgen Receptor Modulators. J. Comb. Chem, 2007, 9(1),107-114.