Thermo Scientific Chemicals

(-)-Cinchonidine, 99% (total base), may cont. up to 5% quinine, Thermo Scientific Chemicals

Catalog number: A18796.22
100 g, Each
Thermo Scientific Chemicals

(-)-Cinchonidine, 99% (total base), may cont. up to 5% quinine, Thermo Scientific Chemicals

Catalog number: A18796.22
100 g, Each
Quantity
Catalog number: A18796.22
also known as A18796-22
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
485-71-2
IUPAC Name
{5-ethenyl-1-azabicyclo[2.2.2]octan-2-yl}(quinolin-4-yl)methanol
Molecular Formula
C19H22N2O
InChI Key
KMPWYEUPVWOPIM-UHFFFAOYNA-N
SMILES
OC(C1CC2CCN1CC2C=C)C1=C2C=CC=CC2=NC=C1
Loss on Drying
≤1.0% (105?C, constant wt.)
Form
Crystals or powder or crystalline powder
Comment
May contain up to 5% Quinine
Optical Rotation
-110? ± 5? (c=1 in ethanol)
Appearance (Color)
White to cream

Description

It finds its uses as an antimalarial compound. It is a lysosomotropic agent, inhibits Plasmodium falciparum, interferes with ferriprotoporphyrin formation and is a weak SERT inhibitor.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It finds its uses as an antimalarial compound. It is a lysosomotropic agent, inhibits Plasmodium falciparum, interferes with ferriprotoporphyrin formation and is a weak SERT inhibitor.

Solubility
Insoluble in water. Soluble in alcohol and chloroform.

Notes
Light Sensitive. Store away from oxidizing agents and light. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text