Thermo Scientific Chemicals

Glycine anhydride, 98%

Catalog number: A18822.14
25 g, Each
Thermo Scientific Chemicals

Glycine anhydride, 98%

Catalog number: A18822.14
25 g, Each
Quantity
Catalog number: A18822.14
also known as A18822-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
106-57-0
IUPAC Name
piperazine-2,5-dione
Molecular Formula
C4H6N2O2
InChI Key
BXRNXXXXHLBUKK-UHFFFAOYSA-N
SMILES
O=C1CNC(=O)CN1
Assay from Supplier's CofA
≥97.5%
Loss on Drying
≤0.5% (105?C, 4h)
Appearance (Color)
White to cream
Form
Crystals or powder or crystalline powder
Identification (FTIR)
Conforms

Description

It can be used as an intermediate in the preparation of various pharmaceutical and biologically active compounds. Also used in the synthesis of dipeptide isosteres by cross-metathesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It can be used as an intermediate in the preparation of various pharmaceutical and biologically active compounds. Also used in the synthesis of dipeptide isosteres by cross-metathesis.

Solubility
Soluble in water (142mg/L).

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text