It can be used as an intermediate in the preparation of various pharmaceutical and biologically active compounds. Also used in the synthesis of dipeptide isosteres by cross-metathesis.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
It can be used as an intermediate in the preparation of various pharmaceutical and biologically active compounds. Also used in the synthesis of dipeptide isosteres by cross-metathesis.
Solubility
Soluble in water (142mg/L).
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
RUO – Research Use Only
A K Szardenings; D Harris; S Lam; L Shi, D Tien; Y Wang; D V Patel; M Navre; D A Campbell. Rational design and combinatorial evaluation of enzyme inhibitor scaffolds: identification of novel inhibitors of matrix metalloproteinases. Journal of Medicinal Chemistry, 1998, 41 (13), 2194-2200.
Da Wang et. al. Diagnostic NH and OH vibrations for oxazolone and diketopiperazine structures: b2 from protonated triglycine. Journal of the American Society for Mass Spectrometry, 2011, 22(7), 1197-1203.