Thermo Scientific Chemicals

Amidosulfonic acid, 98+%, Thermo Scientific Chemicals

Catalog number: A18836.0E
2500 g, Each
Thermo Scientific Chemicals

Amidosulfonic acid, 98+%, Thermo Scientific Chemicals

Catalog number: A18836.0E
2500 g, Each
Quantity
Catalog number: A18836.0E
also known as A18836-0E
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
5329-14-6
IUPAC Name
sulfamic acid
Molecular Formula
H3NO3S
InChI Key
IIACRCGMVDHOTQ-UHFFFAOYSA-N
SMILES
NS(O)(=O)=O
Form
Crystals or powder or crystalline powder
Identification (FTIR)
Conforms
Appearance (Color)
White
Assay (Aqueous acid-base Titration)
≥98.0 to ≤102.0%

Description

Amidosulfonic acid is widely utilized as a green catalyst for the preparation of amide from ketoxime. It finds an application as a titrant in the determination of the burette injection volume and chemical calibration factor. It is an efficient catalyst in the synthesis of polyhydroquinoline derivatives by Hantzsch condensation reaction, deazaoxaflavin, fatty acid methyl and ethyl esters and in the isolation of 5-hydroxymethylfurfural (HMF) from bamboo fiber. It is involved in the determination of silicates in water samples. It acts as a neutralizing agent during the determination of paracetamol by Glynn and Kendal colorimetric method.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Amidosulfonic acid is widely utilized as a green catalyst for the preparation of amide from ketoxime. It finds an application as a titrant in the determination of the burette injection volume and chemical calibration factor. It is an efficient catalyst in the synthesis of polyhydroquinoline derivatives by Hantzsch condensation reaction, deazaoxaflavin, fatty acid methyl and ethyl esters and in the isolation of 5-hydroxymethylfurfural (HMF) from bamboo fiber. It is involved in the determination of silicates in water samples. It acts as a neutralizing agent during the determination of paracetamol by Glynn and Kendal colorimetric method.

Solubility
Slightly soluble in water, dimethyl fluoride and methanol. Insoluble in hydrocarbons.

Notes
Incompatible with chlorine compounds, nitrates, carbonates, strong oxidizing agents and strong bases.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text