cis-2-Nonen-1-ol, 95%, Thermo Scientific Chemicals
cis-2-Nonen-1-ol, 95%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

cis-2-Nonen-1-ol, 95%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
Catalog number A18842.14
also known as A18842-14
Price (USD)/ Each
192.65
Online exclusive
214.00 
Save 21.35 (10%)
-
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (USD)/ Each
192.65
Online exclusive
214.00 
Save 21.35 (10%)
Add to cart
cis-2-Nonen-1-ol, 95%, Thermo Scientific Chemicals
Catalog numberA18842.14
Price (USD)/ Each
192.65
Online exclusive
214.00 
Save 21.35 (10%)
-
Add to cart
Chemical Identifiers
CAS41453-56-9
IUPAC Name(2Z)-non-2-en-1-ol
Molecular FormulaC9H18O
InChI KeyNSSALFVIQPAIQK-FPLPWBNLSA-N
SMILESCCCCCCC=C/CO
View more
SpecificationsSpecification SheetSpecification Sheet
FormLiquid
Identification (FTIR)Conforms
Assay (GC)≥94.0%
Appearance (Color)Clear colorless
Refractive Index1.4470-1.4530 @ 20?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only