Thermo Scientific Chemicals

1,8-Naphthalimide, 98%, Thermo Scientific Chemicals

Catalog number: A18856.18
50 g, Each
Thermo Scientific Chemicals

1,8-Naphthalimide, 98%, Thermo Scientific Chemicals

Catalog number: A18856.18
50 g, Each
Quantity
Catalog number: A18856.18
also known as A18856-18
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
81-83-4
IUPAC Name
3-azatricyclo[7.3.1.0⁵,¹³]trideca-1(13),5,7,9,11-pentaene-2,4-dione
Molecular Formula
C12H7NO2
InChI Key
XJHABGPPCLHLLV-UHFFFAOYSA-N
SMILES
O=C1NC(=O)C2=C3C(C=CC=C13)=CC=C2
Appearance (Color)
Pale cream to cream to pale brown
Form
Powder
Assay (HPLC)
≥97.5%
Identification (FTIR)
Conforms (non-U.S. specification)
GC/MS Analysis
Conforms (U.S. specification)

Description

It is a useful material for utilization in phosphorescence dye doped organic light emitting diodes (oleds), hole blocking and electron-conducting material.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is a useful material for utilization in phosphorescence dye doped organic light emitting diodes (oleds), hole blocking and electron-conducting material.

Solubility
Insoluble in water.

Notes
Store away from and oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text