N,N-Dimethylglycine hydrochloride, 99%, Thermo Scientific Chemicals
N,N-Dimethylglycine hydrochloride, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

N,N-Dimethylglycine hydrochloride, 99%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Catalog number A18892.06
also known as A18892-06
Price (USD)/ Each
38.60
-
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)/ Each
38.60
Add to cart
N,N-Dimethylglycine hydrochloride, 99%, Thermo Scientific Chemicals
Catalog numberA18892.06
Price (USD)/ Each
38.60
-
Add to cart
Chemical Identifiers
CAS2491-06-7
IUPAC Namehydrogen 2-(dimethylamino)acetic acid chloride
Molecular FormulaC4H10ClNO2
InChI KeyFKASAVXZZLJTNX-UHFFFAOYSA-N
SMILES[H+].[Cl-].CN(C)CC(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to pale cream
Assay (Titration ex Chloride)≥98.5% to ≤101.5%
FormCrystals or powder or crystalline powder or lumpy solid
N,N-Dimethylglycine (DMG) is a natural N-methylated glycine that is used in comparative analysis with other N-methylated glycines such as sarcosine and βine. Used in the development of glycine-based ionic liquids and emulsifiers. Also it is potentially useful as a biomarker of protein degradation in COPD patients. DMG is used as a substrate to identify, differentiate and characterize amino acid methyltransferase(s).

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N,N-Dimethylglycine (DMG) is a natural N-methylated glycine that is used in comparative analysis with other N-methylated glycines such as sarcosine and βine. Used in the development of glycine-based ionic liquids and emulsifiers. Also it is potentially useful as a biomarker of protein degradation in COPD patients. DMG is used as a substrate to identify, differentiate and characterize amino acid methyltransferase(s).

Solubility
Soluble in water (almost transparency).

Notes
Hygroscopic, store away from moisture. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
RUO – Research Use Only

General References:

  1. Chen SY; Lai MC; Lai SJ; Lee YCr. Characterization of osmolyte βine synthesizing sarcosine dimethylglycineN-methyltransferase from Methanohalophilus portucalensis. Appl. Microbiol., 2009, 191 (14), 735-743.
  2. Jing-Fang Zhu; Ling He; Lei Zhang; Ming Huang; Guo-Hong Tao. Experimental and theoretical enthalpies of formation of glycine-based sulfate/bisulfate amino acid ionic liquids. Journal of Physical Chemistry B, 2012, 116(1), 113-119.