Thermo Scientific Chemicals

N,N-Dimethylglycine hydrochloride, 99%, Thermo Scientific Chemicals

Catalog number: A18892.14
25 g, Each
Thermo Scientific Chemicals

N,N-Dimethylglycine hydrochloride, 99%, Thermo Scientific Chemicals

Catalog number: A18892.14
25 g, Each
Quantity
Catalog number: A18892.14
also known as A18892-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
2491-06-7
IUPAC Name
hydrogen 2-(dimethylamino)acetic acid chloride
Molecular Formula
C4H10ClNO2
InChI Key
FKASAVXZZLJTNX-UHFFFAOYSA-N
SMILES
[H+].[Cl-].CN(C)CC(O)=O
Appearance (Color)
White to pale cream
Assay (Titration ex Chloride)
≥98.5% to ≤101.5%
Form
Crystals or powder or crystalline powder or lumpy solid

Description

N,N-Dimethylglycine (DMG) is a natural N-methylated glycine that is used in comparative analysis with other N-methylated glycines such as sarcosine and βine. Used in the development of glycine-based ionic liquids and emulsifiers. Also it is potentially useful as a biomarker of protein degradation in COPD patients. DMG is used as a substrate to identify, differentiate and characterize amino acid methyltransferase(s).

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N,N-Dimethylglycine (DMG) is a natural N-methylated glycine that is used in comparative analysis with other N-methylated glycines such as sarcosine and βine. Used in the development of glycine-based ionic liquids and emulsifiers. Also it is potentially useful as a biomarker of protein degradation in COPD patients. DMG is used as a substrate to identify, differentiate and characterize amino acid methyltransferase(s).

Solubility
Soluble in water (almost transparency).

Notes
Hygroscopic, store away from moisture. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text