It is used to produce 6-acetylamino-phthalide. It is also used as a starting material in the reduction reaction.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
It is used to produce 6-acetylamino-phthalide. It is also used as a starting material in the reduction reaction.
Solubility
Soluble in water 1360 mg/L 25°C.
Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
RUO – Research Use Only
General References:
- ProQuest.Design, Synthesis and Polymerization of Highly Branched Pseudodendrimers Through Tandem Reactions and Kinetics Measurements of Pseudodendrimeric Polymerization,1; ProQuest; USA, 2008; 85-103.
- P. Sutter.; C.D. Weis. The synthesis of monosubstituted derivatives of 6-amino-3,3'-bis(p-dimethylaminophenyl)phthalide.Dyes and Pigments.1990,12(4), 287-300.