Thermo Scientific Chemicals

6-Aminophthalide, 95%, Thermo Scientific Chemicals

Catalog number: A18896.88
2.5 g, Each
Thermo Scientific Chemicals

6-Aminophthalide, 95%, Thermo Scientific Chemicals

Catalog number: A18896.88
2.5 g, Each
Quantity
Catalog number: A18896.88
also known as A18896-88
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
57319-65-0
IUPAC Name
6-amino-1,3-dihydro-2-benzofuran-1-one
Molecular Formula
C8H7NO2
InChI Key
ZIJZDNKZJZUROE-UHFFFAOYSA-N
SMILES
NC1=CC=C2COC(=O)C2=C1
Assay (Non-aqueous acid-base Titration)
≥94.0 to ≤106.0%
Form
Powder
Appearance (Color)
Pale cream to cream to yellow to pale brown

Description

It is used to produce 6-acetylamino-phthalide. It is also used as a starting material in the reduction reaction.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used to produce 6-acetylamino-phthalide. It is also used as a starting material in the reduction reaction.

Solubility
Soluble in water 1360 mg/L 25°C.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text