Thermo Scientific Chemicals

4-Ethoxyphenyl isothiocyanate, 98%, Thermo Scientific Chemicals

Catalog number: A18902.14
25 g, Each
Thermo Scientific Chemicals

4-Ethoxyphenyl isothiocyanate, 98%, Thermo Scientific Chemicals

Catalog number: A18902.14
25 g, Each
Quantity
Catalog number: A18902.14
also known as A18902-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
3460-49-9
IUPAC Name
1-ethoxy-4-isothiocyanatobenzene
Molecular Formula
C9H9NOS
InChI Key
REEKOMRZYJXXNR-UHFFFAOYSA-N
SMILES
CCOC1=CC=C(C=C1)N=C=S
Appearance (Color)
White to pale cream to pale yellow
Assay (GC)
≥97.5%
Form
Fused solid
Identification (FTIR)
Conforms
Melting Point (clear melt)
56.0-63.0?C

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text