Thermo Scientific Chemicals

Boc-ON, 98+%, Thermo Scientific Chemicals

Catalog number: A18906.06
5 g, Each
Thermo Scientific Chemicals

Boc-ON, 98+%, Thermo Scientific Chemicals

Catalog number: A18906.06
5 g, Each
Quantity
Catalog number: A18906.06
also known as A18906-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
58632-95-4
IUPAC Name
tert-butyl (Z)-[cyano(phenyl)methylidene]amino carbonate
Molecular Formula
C13H14N2O3
InChI Key
QQWYQAQQADNEIC-RVDMUPIBSA-N
SMILES
CC(C)(C)OC(=O)O\N=C(/C#N)C1=CC=CC=C1
Appearance (Color)
White to pale yellow
Form
Crystals or powder or crystalline powder
Melting Point (clear melt)
84.0-91.0?C
Assay (HPLC)
≥98.0%

Description

Widely used for protection of amino groups in peptide synthesis and multi-step organic synthesis of small molecules.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Widely used for protection of amino groups in peptide synthesis and multi-step organic synthesis of small molecules.

Solubility
Soluble in ethyl acetate, ether and benzene. Insoluble in water.

Notes
Moisture Sensitive, store away from water/moisture. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store under dry inert gas.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text