Chemical Identifiers
CAS118-82-1
IUPAC Name2,6-di-tert-butyl-4-[(3,5-di-tert-butyl-4-hydroxyphenyl)methyl]phenol
Molecular FormulaC29H44O2
InChI KeyMDWVSAYEQPLWMX-UHFFFAOYSA-N
SMILESCC(C)(C)C1=CC(CC2=CC(=C(O)C(=C2)C(C)(C)C)C(C)(C)C)=CC(=C1O)C(C)(C)C
View more