Thermo Scientific Chemicals

Thionin acetate, Thermo Scientific Chemicals

Catalog number: A18912.14
25 g, Each
Thermo Scientific Chemicals

Thionin acetate, Thermo Scientific Chemicals

Catalog number: A18912.14
25 g, Each
Quantity
Catalog number: A18912.14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
78338-22-4
IUPAC Name
3,7-diamino-5λ⁴-phenothiazin-5-ylium
Molecular Formula
C12H10N3S
InChI Key
CBBMGBGDIPJEMI-UHFFFAOYSA-N
SMILES
NC1=CC2=[S+]C3=CC(N)=CC=C3N=C2C=C1
Appearance (Color)
Dark green to black
Form
powder
Assay (unspecified)
Dye content ≥ 85%
Comment
Blue to purple in aqueous solution
lambda-max (UV-Vis)
592 - 602nm in water

Description

Thionin acetate is a strongly staining metachromatic dye that is widely used for biological staining. Thionine can also be used in place of Schiff reagent in quantitative Feulgen staining of DNA. It can also be used to mediate electron transfer in microbial fuel cells. It is also employed as a general nuclear stain for chromatin and mucin; for staining Nissl granules by end-point buffering and for showing malignant cells in frozen biopsy sections; and as a stain for semi-thin sections of plant material for light microscopy.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Thionin acetate is a strongly staining metachromatic dye that is widely used for biological staining. Thionine can also be used in place of Schiff reagent in quantitative Feulgen staining of DNA. It can also be used to mediate electron transfer in microbial fuel cells. It is also employed as a general nuclear stain for chromatin and mucin; for staining Nissl granules by end-point buffering and for showing malignant cells in frozen biopsy sections; and as a stain for semi-thin sections of plant material for light microscopy.

Solubility
Soluble in water (∼2.5 mg/ml at 25°C), and HOAc: H2O (1:1) (1 mg/ml). Insoluble in ethanol.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text