Thermo Scientific Chemicals

2-Acrylamido-2-methylpropanesulfonic acid, 98%, Thermo Scientific Chemicals

Catalog number: A18924.0B
1000 g, Each
Thermo Scientific Chemicals

2-Acrylamido-2-methylpropanesulfonic acid, 98%, Thermo Scientific Chemicals

Catalog number: A18924.0B
1000 g, Each
Quantity
Catalog number: A18924.0B
also known as A18924-0B
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
15214-89-8
IUPAC Name
2-methyl-2-(prop-2-enamido)propane-1-sulfonic acid
Molecular Formula
C7H13NO4S
InChI Key
XHZPRMZZQOIPDS-UHFFFAOYSA-N
SMILES
CC(C)(CS(O)(=O)=O)NC(=O)C=C
Appearance (Color)
White
Form
Powder
Assay (Aqueous acid-base Titration)
≥97.5 to ≤102.5%
Identification (FTIR)
Conforms
Water Content (Karl Fischer Titration)
≤1.0%

Description

2-Acrylamide-2-methylpropanesulfonic acid has good complexion, adsorption, biological activity, surface activity, hydrolysis stability and thermal stability. It can be used in oil chemical, water treatment, synthetic fiber, printing and dyeing, plastics, water absorbing coatings, paper, bio-medical, magnetic materials and cosmetics industries.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Acrylamide-2-methylpropanesulfonic acid has good complexion, adsorption, biological activity, surface activity, hydrolysis stability and thermal stability. It can be used in oil chemical, water treatment, synthetic fiber, printing and dyeing, plastics, water absorbing coatings, paper, bio-medical, magnetic materials and cosmetics industries.

Solubility
Highly soluble in water (1×106 mg/L at 25°C ).

Notes
Hygroscopic. Store away from oxidizing agents and bases. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text