Thermo Scientific Chemicals

1-Bromo-2-methoxynaphthalene, 97%, Thermo Scientific Chemicals

Catalog number: A18926.14
25 g, Each
Thermo Scientific Chemicals

1-Bromo-2-methoxynaphthalene, 97%, Thermo Scientific Chemicals

Catalog number: A18926.14
25 g, Each
Quantity
Catalog number: A18926.14
also known as A18926-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
3401-47-6
IUPAC Name
1-bromo-2-methoxynaphthalene
Molecular Formula
C11H9BrO
InChI Key
XNIGURFWNPLWJM-UHFFFAOYSA-N
SMILES
COC1=CC=C2C=CC=CC2=C1Br
Appearance (Color)
White
Form
Crystals or powder or crystalline powder
Assay (GC)
≥96.0%
Melting Point
79.5-88.5?C

Description

1-Bromo-2-methoxy-naphthalene is a useful synthetic intermediate. It is used as a reagent to synthesize a catalyst for highly enantioselective aziridination of styrene derivatives. It can also be used to prepare biaryls or biheterocycles by palladium-catalyzed Ullmann coupling.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Bromo-2-methoxy-naphthalene is a useful synthetic intermediate. It is used as a reagent to synthesize a catalyst for highly enantioselective aziridination of styrene derivatives. It can also be used to prepare biaryls or biheterocycles by palladium-catalyzed Ullmann coupling.

Solubility
Slightly soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text