Thermo Scientific Chemicals

Bis(4-chlorophenyl) sulfone, 99%, Thermo Scientific Chemicals

Catalog number: A18938.36
500 g, Each
Thermo Scientific Chemicals

Bis(4-chlorophenyl) sulfone, 99%, Thermo Scientific Chemicals

Catalog number: A18938.36
500 g, Each
Quantity
Catalog number: A18938.36
also known as A18938-36
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
80-07-9
IUPAC Name
1-chloro-4-(4-chlorobenzenesulfonyl)benzene
Molecular Formula
C12H8Cl2O2S
InChI Key
GPAPPPVRLPGFEQ-UHFFFAOYSA-N
SMILES
ClC1=CC=C(C=C1)S(=O)(=O)C1=CC=C(Cl)C=C1
Appearance (Color)
White
Form
Crystals or powder or crystalline powder
Assay (GC)
≥98.5%
Melting Point (clear melt)
145.0-152.0?C
Identification (FTIR)
Conforms

Description

Bis(4-chlorophenyl) sulfone is used as a precursor to prepare 4,4'-diamino-diphenyl sulfone and polysulfone plastics. It is also used to prepare rigid and temperature resistant polymers such as PES or Udel through polymerization with bisphenol-A and bisphenol-S. Further, the udel polymer finds applications in plumbing pipes, printer cartridges and automobile fuses.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Bis(4-chlorophenyl) sulfone is used as a precursor to prepare 4,4′-diamino-diphenyl sulfone and polysulfone plastics. It is also used to prepare rigid and temperature resistant polymers such as PES or Udel through polymerization with bisphenol-A and bisphenol-S. Further, the udel polymer finds applications in plumbing pipes, printer cartridges and automobile fuses.

Solubility
Insoluble in water.

Notes
Moisture sensitive. Incompatible with strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text