3-Ethoxybenzaldehyde, 97%, Thermo Scientific Chemicals
3-Ethoxybenzaldehyde, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

3-Ethoxybenzaldehyde, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
2.5 g
Catalog number A18940.09
also known as A18940-09
Price (USD)/ Each
247.00
-
Add to cart
Quantity:
10 g
Request bulk or custom format
Price (USD)/ Each
247.00
Add to cart
3-Ethoxybenzaldehyde, 97%, Thermo Scientific Chemicals
Catalog numberA18940.09
Price (USD)/ Each
247.00
-
Add to cart
Chemical Identifiers
CAS22924-15-8
IUPAC Name3-ethoxybenzaldehyde
Molecular FormulaC9H10O2
InChI KeyQZMGMXBYJZVAJN-UHFFFAOYSA-N
SMILESCCOC1=CC=CC(C=O)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (GC)≥96.0%
FormLiquid
Refractive Index1.5365-1.5415 @ 20?C
Appearance (Color)Clear colorless
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only