Thermo Scientific Chemicals

cis-1,4-Dichloro-2-butene, 95%, Thermo Scientific Chemicals

Catalog number: A18956.14
25 g, Each
Thermo Scientific Chemicals

cis-1,4-Dichloro-2-butene, 95%, Thermo Scientific Chemicals

Catalog number: A18956.14
25 g, Each
Quantity
Catalog number: A18956.14
also known as A18956-14
Price (USD)
Have Questions?

Chemical Identifiers

CAS
1476-11-5
IUPAC Name
(2Z)-1,4-dichlorobut-2-ene
Molecular Formula
C4H6Cl2
InChI Key
FQDIANVAWVHZIR-UPHRSURJSA-N
SMILES
ClCC=C/CCl
Appearance (Color)
Clear colorless to yellow
Assay (GC)
≥94.0% (UK sourced material)
Refractive Index
1.4865-1.4925 @ 20?C (UK sourced material)
Form
Liquid
Comment
Material sourced in UK and US

Description

It is employed as an intermediate for the production of other chemicals such as in the production of chloroprene. It is also used as a starting material in the production of adiponitrile, butane-1,4-diol, and tetrahydrofuran. It is also used in the preparation of functionalized 3,5-disubstituted cyclopent-2-enones.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is employed as an intermediate for the production of other chemicals such as in the production of chloroprene. It is also used as a starting material in the production of adiponitrile, butane-1,4-diol, and tetrahydrofuran. It is also used in the preparation of functionalized 3,5-disubstituted cyclopent-2-enones.

Solubility
Not miscible or difficult to mix in water. Soluble in alcohol, ether, acetone and benzene.

Notes
Moisture & Light Sensitive. Store away from oxidizing agents, heat, water/moisture and light. Store under inert gas. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Keep it refrigerated.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text