3,3-Dimethylallyl acetate, 98%, stab. with 0.1% alpha tocopherol, Thermo Scientific Chemicals
3,3-Dimethylallyl acetate, 98%, stab. with 0.1% alpha tocopherol, Thermo Scientific Chemicals
Thermo Scientific Chemicals

3,3-Dimethylallyl acetate, 98%, stab. with 0.1% alpha tocopherol, Thermo Scientific Chemicals

Have Questions?
Quantity:
50 g
Catalog number A18958.18
also known as A18958-18
Price (USD)/ Each
48.10
-
Add to cart
Quantity:
50 g
Request bulk or custom format
Price (USD)/ Each
48.10
Add to cart
3,3-Dimethylallyl acetate, 98%, stab. with 0.1% alpha tocopherol, Thermo Scientific Chemicals
Catalog numberA18958.18
Price (USD)/ Each
48.10
-
Add to cart
Chemical Identifiers
CAS1191-16-8
IUPAC Name3-methylbut-2-en-1-yl acetate
Molecular FormulaC7H12O2
InChI KeyXXIKYCPRDXIMQM-UHFFFAOYSA-N
SMILESCC(C)=CCOC(C)=O
View more
SpecificationsSpecification SheetSpecification Sheet
FormLiquid
Refractive Index1.4275-1.4315 @ 20?C
Identification (FTIR)Conforms
Appearance (Color)Clear colorless
Assay (GC)≥97.5%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only