Thermo Scientific Chemicals

5-Iodouracil, 97%, Thermo Scientific Chemicals

Catalog number: A18994.18
50 g, Each
Thermo Scientific Chemicals

5-Iodouracil, 97%, Thermo Scientific Chemicals

Catalog number: A18994.18
50 g, Each
Quantity
Catalog number: A18994.18
also known as A18994-18
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
696-07-1
IUPAC Name
5-iodo-1,2,3,4-tetrahydropyrimidine-2,4-dione
Molecular Formula
C4H3IN2O2
InChI Key
KSNXJLQDQOIRIP-UHFFFAOYSA-N
SMILES
IC1=CNC(=O)NC1=O
Form
Powder
Assay (Aqueous acid-base Titration)
≥96.0 to ≤104.0%
Appearance (Color)
White to pale cream

Description

It is used as an intermediate. It has an antitumor activity.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used as an intermediate. It has an antitumor activity.

Solubility
Very faint turbidity in NH3aq. Soluble in 1M NaOH.

Notes
Light Sensitive. Store away from oxidizing agents and light. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text