Thermo Scientific Chemicals

3,4,5-Trimethoxyphenylacetone, 98%, Thermo Scientific Chemicals

Catalog number: A19006.14
25 g, Each
Thermo Scientific Chemicals

3,4,5-Trimethoxyphenylacetone, 98%, Thermo Scientific Chemicals

Catalog number: A19006.14
25 g, Each
Quantity
Catalog number: A19006.14
also known as A19006-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
16603-18-2
IUPAC Name
1-(3,4,5-trimethoxyphenyl)propan-2-one
Molecular Formula
C12H16O4
InChI Key
WDNDMVGQGRDIBC-UHFFFAOYSA-N
SMILES
COC1=CC(CC(C)=O)=CC(OC)=C1OC
Identification (FTIR)
Conforms
Form
Crystals or powder or crystalline powder
Appearance (Color)
White to pale cream to cream to brown
Assay (GC)
≥97.5%
Melting Point (clear melt)
53.0-64.0?C

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text