Thermo Scientific Chemicals

5-Methylhexanoic acid, 98%, Thermo Scientific Chemicals

Catalog number: A19012.09
10 g, Each
Thermo Scientific Chemicals

5-Methylhexanoic acid, 98%, Thermo Scientific Chemicals

Catalog number: A19012.09
10 g, Each
Quantity
Catalog number: A19012.09
also known as A19012-09
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
628-46-6
Appearance (Color)
Clear colorless
Assay (Aqueous acid-base Titration)
≥97.5 to ≤102.5%
Refractive Index
1.4200-1.4240 @ 20?C
Form
Liquid
Assay (GC)
≥97.5%

Description

Ion-exclusion chromatographic separations of C1-C6 aliphatic carboxylic acids on a sulfonated styrene-divinylbenzene co-polymer resin column, high sensitive conductimetric detection is achieved with 5-methylhexanoic acid as eluent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Ion-exclusion chromatographic separations of C1-C6 aliphatic carboxylic acids on a sulfonated styrene-divinylbenzene co-polymer resin column, high sensitive conductimetric detection is achieved with 5-methylhexanoic acid as eluent.

Solubility
Slightly soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, bases.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text