Thermo Scientific Chemicals

2-Benzyloxyphenol, 98%, Thermo Scientific Chemicals

Catalog number: A19020.14
25 g, Each
Thermo Scientific Chemicals

2-Benzyloxyphenol, 98%, Thermo Scientific Chemicals

Catalog number: A19020.14
25 g, Each
Quantity
Catalog number: A19020.14
also known as A19020-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
6272-38-4
IUPAC Name
2-(benzyloxy)phenol
Molecular Formula
C13H12O2
InChI Key
CCZCXFHJMKINPE-UHFFFAOYSA-N
SMILES
OC1=CC=CC=C1OCC1=CC=CC=C1
Form
Liquid or viscous liquid
Assay (GC)
≥97.5%
Appearance (Color)
Clear colorless to pale brown or pale pink to red
Refractive Index
1.5885-1.5925 @ 20?C

Description

2-Benzyloxyphenol is used in the synthesis of 2-(benzyloxy)hydroquinone and also used to prepare sequential polypeptides.It is used as reagent for synthesis of multidentate chelating ligands. It is also used in chemicals, pharmaceutical research and also acts a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Benzyloxyphenol is used in the synthesis of 2-(benzyloxy)hydroquinone and also used to prepare sequential polypeptides.It is used as reagent for synthesis of multidentate chelating ligands. It is also used in chemicals, pharmaceutical research and also acts a pharmaceutical intermediate.

Solubility
Soluble in alcohol, benzene, and diethyl ether. Insoluble in water.

Notes
Incompatible with strong oxidizing agents and strong acids.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text