Thermo Scientific Chemicals

(R)-(-)-2-Phenylglycinol, 98%, Thermo Scientific Chemicals

Catalog number: A19030.14
25 g, Each
Thermo Scientific Chemicals

(R)-(-)-2-Phenylglycinol, 98%, Thermo Scientific Chemicals

Catalog number: A19030.14
25 g, Each
Quantity
Catalog number: A19030.14
also known as A19030-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
56613-80-0
IUPAC Name
(1S)-2-hydroxy-1-phenylethan-1-aminium
Molecular Formula
C8H12NO
InChI Key
IJXJGQCXFSSHNL-MRVPVSSYSA-O
SMILES
[NH3+][C@H](CO)C1=CC=CC=C1
Appearance (Color)
White to cream to yellow
Assay (HPLC)
≥97.5%
Optical Rotation
-30.5 ± 2.5? (C=1.0 in 1N HCl)
Form
Crystals or powder or crystalline powder
Melting Point (clear melt)
74.0-81.0?C

Description

Amino alcohol used to prepare a chiral imine or oxazolidine from ethyl trifluoropyruvate. These intermediates were then employed in a synthesis of both enantiomers of α-trifluoromethylproline. Chiral β-amino alcohol used as a synthetic building block.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Amino alcohol used to prepare a chiral imine or oxazolidine from ethyl trifluoropyruvate. These intermediates were then employed in a synthesis of both enantiomers of α-trifluoromethylproline. Chiral β-amino alcohol used as a synthetic building block.

Solubility
Slightly soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text