2-Tridecanone, 98+%, Thermo Scientific Chemicals
2-Tridecanone, 98+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

2-Tridecanone, 98+%, Thermo Scientific Chemicals

Have Questions?
Quantity:
100 g
Catalog number A19042.22
also known as A19042-22
Price (USD)/ Each
113.00
-
Add to cart
Quantity:
100 g
Request bulk or custom format
Price (USD)/ Each
113.00
Add to cart
2-Tridecanone, 98+%, Thermo Scientific Chemicals
Catalog numberA19042.22
Price (USD)/ Each
113.00
-
Add to cart
Chemical Identifiers
CAS593-08-8
IUPAC Nametridecan-2-one
Molecular FormulaC13H26O
InChI KeyCYIFVRUOHKNECG-UHFFFAOYSA-N
SMILESCCCCCCCCCCCC(C)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to black
FormCrystals or powder or crystalline powder or lumps or fused solid
Assay (GC)≥98.0%
Identification (FTIR)Conforms
Melting Point (clear melt)26.0-32.0?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only