Thermo Scientific Chemicals

2,4,6-Trichloroaniline, 98+%, Thermo Scientific Chemicals

Catalog number: A19044.22
100 g, Each
Thermo Scientific Chemicals

2,4,6-Trichloroaniline, 98+%, Thermo Scientific Chemicals

Catalog number: A19044.22
100 g, Each
Quantity
Catalog number: A19044.22
also known as A19044-22
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
634-93-5
IUPAC Name
2,4,6-trichloroaniline
Molecular Formula
C6H4Cl3N
InChI Key
NATVSFWWYVJTAZ-UHFFFAOYSA-N
SMILES
NC1=C(Cl)C=C(Cl)C=C1Cl
Appearance (Color)
White to pale cream
Form
Crystals or powder or crystalline powder
Assay (GC)
≥98.0%
Identification (FTIR)
Conforms
Melting Point (clear melt)
74.0-81.0?C

Description

Tetrachloronitrobenzene was prepared from 2:4:6-trichloroaniline,which was converted by a Sandmeyer reaction into 1:2:3:5- tetrachlorobenzene.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Tetrachloronitrobenzene was prepared from 2:4:6-trichloroaniline,which was converted by a Sandmeyer reaction into 1:2:3:5- tetrachlorobenzene.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text