2,4,5-Trimethylthiazole, 98%, Thermo Scientific Chemicals
2,4,5-Trimethylthiazole, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

2,4,5-Trimethylthiazole, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
Catalog number A19046.14
also known as A19046-14
Price (USD)/ Each
160.00
-
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (USD)/ Each
160.00
Add to cart
2,4,5-Trimethylthiazole, 98%, Thermo Scientific Chemicals
Catalog numberA19046.14
Price (USD)/ Each
160.00
-
Add to cart
Chemical Identifiers
CAS13623-11-5
IUPAC Nametrimethyl-1,3-thiazole
Molecular FormulaC6H9NS
InChI KeyBAMPVSWRQZNDQC-UHFFFAOYSA-N
SMILESCC1=NC(C)=C(C)S1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear pale yellow to yellow
Assay (GC)≥97.5%
FormLiquid
Refractive Index1.5060-1.5100 @ 20?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only